Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007631.2 | NP_031657.1 | 2 | 569 | ||
GenBank mRNA | AK005352.1 | - | 2 | 263 | ||
AK017834.1 | - | 2 | 533 | |||
AK053348.1 | - | 2 | 477 | |||
AK149905.1 | - | 2 | 476 | |||
AK164997.1 | - | 2 | 514 | |||
AK170833.1 | - | 2 | 450 | |||
BC044841.1 | - | 2 | 472 | 3155470 | ||
BC060172.1 | - | 2 | 464 | |||
EU600170.1 | - | 2 | 569 | |||
M64403.1 | - | 2 | 524 | |||
S78355.1 | - | 2 | 474 |