Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007634.4 | NP_031660.3 | 3 | 362 | ||
GenBank mRNA | AK030298.1 | - | 301 | |||
AK031613.1 | - | 3 | 302 | |||
AK032961.1 | - | 3 | 288 | |||
AK082485.1 | - | 3 | 330 | |||
AK085183.1 | - | 3 | 299 | |||
AK159594.1 | - | 3 | 363 | |||
AK162447.1 | - | 3 | 302 | |||
AK169263.1 | - | 3 | 313 | |||
AK211279.1 | - | 2 | 200 | |||
BC021658.1 | - | 3 | 282 | |||
BC037662.1 | - | 3 | 314 | 5353169 | ||
U20612.1 | - | 3 | 300 | |||
U20636.1 | - | 3 | 417 |