Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007637.2 | NP_031663.1 | 7, 8 | 1089 | ||
GenBank mRNA | AK077753.1 | - | 7, 8 | 1047 | ||
AK088184.1 | - | 7, 8 | 1023 | |||
AK129056.1 | - | 7, 8 | 995 | |||
AK133857.1 | - | 7, 8 | 1047 | |||
AK136493.1 | - | 7, 8 | 1041 | |||
AK145445.1 | - | 7, 8 | 1058 | |||
AK146405.1 | - | 7, 8 | 1061 | |||
AK146425.1 | - | 7, 8 | 1041 | |||
AK150390.1 | - | 7, 8 | 1008 | |||
AK153911.1 | - | 7, 8 | 1065 | |||
AK167895.1 | - | 7, 8 | 1030 | |||
BC092233.1 | - | 7, 8 | 1059 | |||
BC094427.1 | - | 7, 8 | 1036 | 30537966 | ||
Z31555.1 | - | 7, 8 | 1089 |