Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009840.3 | NP_033970.3 | 2, 3 | 227 | ||
GenBank mRNA | AK010882.1 | - | 2, 3 | 228 | ||
AK075711.1 | - | 2, 3 | 208 | |||
AK075880.1 | - | 2, 3 | 216 | |||
AK145661.1 | - | 2, 3 | 211 | |||
AK145754.1 | - | 2, 3 | 210 | |||
AK145918.1 | - | 2, 3 | 229 | |||
AK147038.1 | - | 2, 3 | 230 | |||
AK159969.1 | - | 2, 3 | 219 | |||
AK168465.1 | - | 2, 3 | 199 | |||
AK172867.1 | - | 2, 3 | 198 | |||
AK184650.1 | - | 1, 2 | 27 | |||
BC009007.1 | - | 2, 3 | 185 | 3582639 | ||
Z37164.1 | - | 2, 3 | 231 |