Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009840.3 | NP_033970.3 | 8, 9 | 1020 | ||
GenBank mRNA | AK010882.1 | - | 8, 9 | 1021 | ||
AK014005.1 | - | 1, 2 | 86 | |||
AK075711.1 | - | 8, 9 | 1001 | |||
AK075880.1 | - | 8, 9 | 1009 | |||
AK145661.1 | - | 8, 9 | 1004 | |||
AK145754.1 | - | 8, 9 | 1003 | |||
AK145918.1 | - | 8, 9 | 1022 | |||
AK147038.1 | - | 8, 9 | 1023 | |||
AK159969.1 | - | 8, 9 | 1012 | |||
AK168465.1 | - | 8, 9 | 993 | |||
AK172867.1 | - | 8, 9 | 991 | |||
AK218945.1 | - | 1, 2 | 47 | |||
BC009007.1 | - | 8, 9 | 978 | 3582639 | ||
Z37164.1 | - | 8, 9 | 1024 |