Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026214.2 | NP_001021385.1 | 13, 14 | 1282 | ||
NM_001286049.1 | NP_001272978.1 | 12, 13 | 1257 | |||
NM_001286058.1 | NP_001272987.1 | 11, 12 | 1184 | |||
NM_007647.3 | NP_031673.2 | 12, 13 | 1209 | |||
GenBank mRNA | AF006482.1 | - | 12, 13 | 1217 | ||
AF136571.1 | - | 12, 13 | 1243 | |||
AJ238636.1 | - | 12, 13 | 1245 | |||
AK002618.1 | - | 12, 13 | 1182 | |||
AK031581.1 | - | 13, 14 | 1262 | |||
AK036641.1 | - | 13, 14 | 1259 | |||
AK037736.1 | - | 12, 13 | 1178 | |||
AK045828.1 | - | 12, 13 | 1195 | |||
AK079267.1 | - | 11, 12 | 1106 | |||
AK080265.1 | - | 13, 14 | 1269 | |||
AK081435.1 | - | 13, 14 | 1247 | |||
AK088455.1 | - | 12, 13 | 1238 | |||
AK160950.1 | - | 10, 11 | 1103 | |||
AK163688.1 | - | 12, 13 | 1183 | |||
BC015247.1 | - | 11, 12 | 1167 | 4242560 |