Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133655.2 | NP_598416.1 | 2 | 393 | ||
GenBank mRNA | AK136370.1 | - | 2 | 357 | ||
AK166521.1 | - | 2 | 359 | |||
AK170741.1 | - | 2 | 358 | |||
AK190176.1 | - | 2 | 274 | |||
BC011433.1 | - | 2 | 348 | 2811801 | ||
BY252638.1 | - | 2 | 394 | |||
X59047.1 | - | 2 | 113 |