Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016756.4 | NP_058036.1 | 2 | 376 | ||
NM_183417.3 | NP_904326.1 | 2 | 376 | |||
GenBank mRNA | AJ223732.1 | - | 2 | 278 | ||
AK036589.1 | - | 1 | 1037 | |||
AK042462.1 | - | 1 | 1037 | |||
AK146845.1 | - | 2 | 356 | |||
AK148017.1 | - | 2 | 315 | |||
AK151979.1 | - | 2 | 326 | |||
AK152922.1 | - | 2 | 329 | |||
AK153983.1 | - | 2 | 328 | |||
AK154997.1 | - | 2 | 328 | |||
AK181235.1 | - | 1 | 47 | |||
AK186022.1 | - | 1 | 47 | |||
AK186779.1 | - | 1 | 47 | |||
AK187629.1 | - | 2 | 112 | |||
AK189612.1 | - | 2 | 112 | |||
AK210959.1 | - | 1 | 47 | |||
AK219393.1 | - | 1 | 47 | |||
BC005654.1 | - | 2 | 337 | 3709730 | ||
BC026145.1 | - | 1 | 1082 | |||
CN705407.1 | - | 2 | 376 | |||
U63337.1 | - | 2 | 304 |