Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007718.3 | NP_031744.3 | 1 | 84 | ||
GenBank mRNA | BC127143.1 | - | 1 | 84 | 40051964 | |
BC128466.1 | - | 1 | 84 | 40051965 |