Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301713.1 | NP_001288642.1 | 2 | 687 | ||
NM_007719.2 | NP_031745.2 | 3 | 737 | |||
GenBank mRNA | AK037965.1 | - | 3 | 738 | ||
AK154349.1 | - | 3 | 738 | |||
AK154736.1 | - | 3 | 738 | |||
AK154849.1 | - | 3 | 738 |