Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290825.1 | NP_001277754.1 | 49 | 3942 | ||
NM_007732.2 | NP_031758.2 | 48 | 3831 | |||
GenBank mRNA | BC003208.1 | - | 7 | 703 | ||
BC125031.1 | - | 49 | 3942 | 40086689 | ||
BC125032.1 | - | 49 | 3941 | 40086691 | ||
CB321186.1 | - | 4 | 402 | |||
L08407.1 | - | 48 | 3883 |