Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109991.1 | NP_001103461.1 | 2 | 1104 | ||
NM_009929.3 | NP_034059.2 | 3 | 520 | |||
GenBank mRNA | AK031798.1 | - | 3 | 508 | ||
BC043697.1 | - | 2 | 1104 | |||
BC062931.1 | - | 3 | 486 | |||
BC064817.1 | - | 3 | 491 | 6332464 | ||
BC066080.1 | - | 3 | 486 | 30355615 | ||
L16898.1 | - | 3 | 459 | |||
L22545.1 | - | 1 | 358 |