Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007735.2 | NP_031761.1 | 44 | 4446 | ||
GenBank mRNA | AF169388.1 | - | 44 | 4389 | ||
BC038814.1 | - | 4 | 206 |