Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007758.2 | NP_031784.1 | 11 | 2123 | ||
GenBank mRNA | AK003072.2 | - | 11 | 2123 | ||
BC109344.1 | - | 11 | 2056 | 40054528 | ||
M29281.1 | - | 6 | 1127 | |||
M35684.1 | - | 10 | 1991 | |||
M61132.1 | - | 10 | 1958 |