Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009983.2 | NP_034113.1 | 2 | 147 | ||
GenBank mRNA | AK089294.1 | - | 2 | 105 | ||
AK150363.1 | - | 2 | 108 | |||
AK150364.1 | - | 2 | 105 | |||
AK150579.1 | - | 2 | 105 | |||
AK150622.1 | - | 2 | 105 | |||
AK150636.1 | - | 2 | 105 | |||
AK150694.1 | - | 2 | 105 | |||
AK150768.1 | - | 2 | 105 | |||
AK150946.1 | - | 2 | 157 | |||
AK151256.1 | - | 2 | 105 | |||
AK151275.1 | - | 2 | 105 | |||
AK151276.1 | - | 2 | 105 | |||
AK151374.1 | - | 2 | 105 | |||
AK151829.1 | - | 2 | 105 | |||
AK151962.1 | - | 2 | 108 | |||
AK152036.1 | - | 2 | 118 | |||
AK152045.1 | - | 2 | 158 | |||
AK152168.1 | - | 2 | 105 | |||
AK152187.1 | - | 2 | 105 | |||
AK152188.1 | - | 2 | 105 | |||
AK152406.1 | - | 2 | 108 | |||
AK152458.1 | - | 2 | 105 | |||
AK152567.1 | - | 2 | 105 | |||
AK152593.1 | - | 2 | 135 | |||
AK152638.1 | - | 2 | 135 | |||
AK152680.1 | - | 2 | 105 | |||
AK152828.1 | - | 2 | 105 | |||
AK152881.1 | - | 2 | 105 | |||
AK152892.1 | - | 2 | 105 | |||
AK153191.1 | - | 2 | 105 | |||
AK153291.1 | - | 2 | 105 | |||
AK159097.1 | - | 2 | 105 | |||
AK159209.1 | - | 2 | 105 | |||
AK159440.1 | - | 2 | 158 | |||
AK159490.1 | - | 2 | 105 | |||
AK159512.1 | - | 2 | 147 | |||
AK159580.1 | - | 2 | 105 | |||
AK159743.1 | - | 2 | 109 | |||
AK159896.1 | - | 2 | 105 | |||
AK159970.1 | - | 2 | 158 | |||
AK165227.1 | - | 2 | 174 | |||
AK166864.1 | - | 2 | 118 | |||
AK166938.1 | - | 2 | 152 | |||
AK167143.1 | - | 2 | 143 | |||
AK167366.1 | - | 2 | 177 | |||
AK167698.1 | - | 2 | 105 | |||
AK167750.1 | - | 2 | 118 | |||
AK167867.1 | - | 2 | 106 | |||
AK169139.1 | - | 2 | 158 | |||
AK171195.1 | - | 2 | 106 | |||
AK172583.1 | - | 2 | 105 | |||
BC048900.1 | - | 2 | 155 | |||
BC053937.1 | - | 2 | 155 | |||
BC053940.1 | - | 2 | 155 | |||
BC054758.1 | - | 2 | 177 | 6413315 | ||
BC057931.1 | - | 2 | 138 | 5253319 | ||
X52886.1 | - | 2 | 147 | |||
X53337.1 | - | 2 | 87 |