Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007802.4 | NP_031828.2 | 2, 3 | 258 | ||
GenBank mRNA | AK003425.1 | - | 2, 3 | 183 | ||
AK132648.1 | - | 2, 3 | 188 | |||
BC046320.1 | - | 2, 3 | 212 | 6390854 | ||
X94444.1 | - | 1, 2 | 130 |