Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010008.4 | NP_034138.3 | 1 | 294 | ||
GenBank mRNA | AK033614.1 | - | 1 | 297 | ||
AK038636.1 | - | 1 | 201 | |||
AK045421.1 | - | 1 | 284 | |||
AK078893.1 | - | 1 | 120 | |||
AK084025.1 | - | 1 | 304 | |||
AK165663.1 | - | 1 | 139 | |||
AK210777.1 | - | 1 | 80 | |||
BC050832.1 | - | 1 | 288 | 6514145 | ||
U62295.1 | - | 1 | 341 |