Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007861.5 | NP_031887.2 | 2, 3 | 253 | ||
GenBank mRNA | AK014092.1 | - | 2, 3 | 196 | ||
AK117104.1 | - | 2, 3 | 197 | |||
AK135807.1 | - | 2, 3 | 196 | |||
AK136193.1 | - | 2, 3 | 197 | |||
AK153399.1 | - | 2, 3 | 197 | |||
AK167885.1 | - | 2, 3 | 197 | |||
AK168157.1 | - | 2, 3 | 218 | |||
AK168875.1 | - | 2, 3 | 138 | |||
BC003368.1 | - | 2, 3 | 180 | 3589344 | ||
BY251665.1 | - | 2, 3 | 220 | |||
BY282701.1 | - | 2, 3 | 256 | |||
U73445.1 | - | 2, 3 | 230 |