Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028953.2 | NP_083229.1 | 16 | 1888 | ||
GenBank mRNA | AF417579.1 | - | 16 | 1888 | ||
AK016832.1 | - | 2 | 239 | |||
AK077146.1 | - | 2 | 248 | |||
AY263155.1 | - | 2 | 162 |