Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007912.4 | NP_031938.1 | 5 | 894 | ||
NM_207655.2 | NP_997538.1 | 5 | 894 | |||
GenBank mRNA | AF124513.1 | - | 5 | 828 | ||
AF275367.1 | - | 5 | 837 | |||
AK004883.1 | - | 5 | 895 | |||
AK004911.1 | - | 5 | 869 | |||
AK004944.1 | - | 5 | 888 | |||
AK134262.1 | - | 5 | 868 | |||
BC023729.1 | - | 5 | 865 | 5351807 | ||
U03425.1 | - | 5 | 664 | |||
X59698.1 | - | 5 | 773 | |||
X78987.1 | - | 5 | 673 |