Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010121.2 | NP_034251.2 | 16, 17 | 3346 | ||
GenBank mRNA | AF076681.1 | - | 16, 17 | 3338 | ||
AK003226.1 | - | 4, 5 | 842 | |||
BC054809.1 | - | 16, 17 | 3358 | 6839221 | ||
CA316032.1 | - | 3, 4 | 193 |