Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007925.3 | NP_031951.2 | 21 | 1349 | ||
GenBank mRNA | AK041860.1 | - | 21 | 1350 | ||
AK080277.1 | - | 21 | 1376 | |||
BC034057.1 | - | 9 | 886 | |||
BC051649.1 | - | 21 | 1418 | 5346255 | ||
U08210.1 | - | 21 | 1336 |