Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033711.1 | NP_001028883.1 | 2 | 1593 | ||
NM_010161.3 | NP_034291.1 | 2 | 780 | |||
GenBank mRNA | AK044098.1 | - | 2 | 656 | ||
AK046407.1 | - | 2 | 731 | |||
AK160302.1 | - | 2 | 697 | |||
AK162958.1 | - | 656 | ||||
AK170527.1 | - | 2 | 752 | |||
AK171332.1 | - | 1 | 3137 | |||
AK171594.1 | - | 2 | 698 | |||
BC038124.1 | - | 2 | 737 | 1396641 | ||
M34896.1 | - | 2 | 710 | |||
M34897.1 | - | 1 | 1106 |