Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080462.2 | NP_536710.1 | 2 | 111 | ||
GenBank mRNA | AB070524.1 | - | 2 | 107 | ||
AB444950.1 | - | 1 | 41 | |||
AB444951.1 | - | 1 | 41 | |||
AF297037.1 | - | 2 | 111 | |||
AK050129.1 | - | 2 | 134 | |||
AK160131.1 | - | 1 | 55 | |||
BC033928.1 | - | 2 | 103 | 1546890 |