Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207223.1 | NP_997106.1 | 3, 4 | 748 | ||
GenBank mRNA | AK147505.1 | - | 3, 4 | 802 | ||
AK159055.1 | - | 1, 2 | 23 | |||
BC025556.1 | - | 3, 4 | 322 | |||
BC067016.1 | - | 3, 4 | 748 | 6827624 | ||
BC098196.1 | - | 3, 4 | 324 | 6847993 |