Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010166.3 | NP_034296.2 | 17 | 1626 | ||
NM_210071.2 | NP_997592.1 | 16 | 1587 | |||
NM_211357.2 | NP_997596.1 | 14 | 1406 | |||
GenBank mRNA | AJ007996.1 | - | 4 | 319 | ||
AK144653.1 | - | 16 | 1578 | |||
BC052860.1 | - | 15 | 1390 | |||
BC063259.1 | - | 16 | 1515 | 6406090 | ||
U61112.1 | - | 13 | 1291 | |||
U81604.1 | - | 17 | 1594 |