Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159521.2 | NP_001152993.1 | 4 | 808 | ||
NM_001284506.1 | NP_001271435.1 | 3 | 722 | |||
NM_138749.3 | NP_620088.2 | 4 | 772 | |||
GenBank mRNA | AK147538.1 | - | 4 | 773 | ||
AK154627.1 | - | 4 | 724 | |||
BC157960.1 | - | 4 | 743 | 9088132 | ||
BC157961.1 | - | 4 | 743 | 9088133 | ||
BC158086.1 | - | 4 | 709 | 9088174 | ||
BC172162.1 | - | 4 | 709 | 9054832 |