Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031164.2 | NP_112441.2 | 11 | 1967 | ||
GenBank mRNA | AK149397.1 | - | 11 | 1954 | ||
BC030166.1 | - | 11 | 1952 | 5051604 | ||
D10071.1 | - | 11 | 1947 |