Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255993.1 | NP_001242922.1 | 6 | 1754 | ||
NM_001255994.1 | NP_001242923.1 | 5 | 1607 | |||
NM_080562.5 | NP_542129.2 | 5 | 1612 | |||
GenBank mRNA | AF360997.1 | - | 5 | 1542 | ||
AK034114.1 | - | 5 | 1561 | |||
AK049276.1 | - | 4 | 1378 | |||
AK158052.1 | - | 6 | 1712 | |||
AK170100.1 | - | 5 | 1584 | |||
AK171070.1 | - | 6 | 1720 | |||
AK177084.1 | - | 2 | 280 | |||
BC025068.1 | - | 4 | 1474 | 4242459 |