Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153055.3 | NP_694695.3 | 4 | 645 | ||
GenBank mRNA | AK036629.1 | - | 4 | 340 | ||
AK049737.1 | - | 4 | 336 | |||
AY024346.1 | - | 4 | 442 | |||
AY238936.1 | - | 4 | 415 | |||
BC019366.1 | - | 4 | 409 | 5066908 | ||
BC031846.1 | - | 4 | 409 | 5066822 | ||
BC059816.1 | - | 4 | 644 | |||
BC080810.1 | - | 4 | 526 |