Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007993.2 | NP_032019.2 | 44 | 5676 | ||
GenBank mRNA | L29454.1 | - | 7763 | |||
U22493.1 | - | 44 | 5609 |