Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271616.1 | NP_001258545.1 | 3 | 776 | ||
NM_008009.4 | NP_032035.2 | 2 | 726 | |||
GenBank mRNA | AF065441.1 | - | 1 | 619 | ||
AK146625.1 | - | 2 | 691 | |||
BC065774.1 | - | 2 | 689 | 3975150 | ||
CB524251.1 | - | 1 | 37 | |||
DV059653.1 | - | 3 | 758 |