Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163215.2 | NP_001156687.1 | 9 | 1561 | ||
NM_001163216.2 | NP_001156688.1 | 7 | 1276 | |||
NM_001163217.2 | NP_001156689.1 | 8 | 1336 | |||
NM_001205270.1 | NP_001192199.1 | 9 | 1535 | |||
NM_008010.5 | NP_032036.2 | 9 | 1535 | |||
GenBank mRNA | AF024638.1 | - | 7 | 1184 | ||
AK143592.1 | - | 9 | 1468 | |||
AK163495.1 | - | 7 | 1114 | |||
BC053056.1 | - | 9 | 1445 | 5708838 | ||
BU053488.1 | - | 4 | 551 | |||
M81342.1 | - | 9 | 1453 | |||
S56291.1 | - | 8 | 1280 | |||
X58255.1 | - | 9 | 1439 |