Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164717.1 | NP_001158189.1 | 14 | 2352 | ||
NM_008018.4 | NP_032044.2 | 15 | 2436 | |||
GenBank mRNA | AJ007012.1 | - | 15 | 2436 | ||
BC118022.1 | - | 14 | 2188 | 40097448 |