Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293621.1 | NP_001280550.1 | 21 | 2547 | ||
NM_008060.2 | NP_032086.1 | 22 | 2613 | |||
GenBank mRNA | AK017873.1 | - | 9 | 953 | ||
AK028655.1 | - | 21 | 2517 | |||
AK030722.1 | - | 21 | 2516 | |||
AK081915.1 | - | 21 | 2509 | |||
AK122201.1 | - | 16 | 2743 | |||
AK166150.1 | - | 21 | 2486 | |||
BC020386.1 | - | 14 | 1763 | |||
BC035423.1 | - | 12 | 1363 | |||
BC094437.1 | - | 21 | 2497 | 30682580 | ||
BC117888.1 | - | 21 | 2490 | 40096413 | ||
BC117889.1 | - | 21 | 2493 | 40096414 | ||
BC128069.1 | - | 22 | 2557 | 40096428 | ||
U92793.1 | - | 22 | 2559 |