Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293630.1 | NP_001280559.1 | 4 | 1107 | ||
NM_001293631.1 | NP_001280560.1 | 4 | 820 | |||
NM_008063.2 | NP_032089.2 | 3 | 847 | |||
GenBank mRNA | AK003620.1 | - | 3 | 670 | ||
AK147849.1 | - | 4 | 777 | |||
AK155339.1 | - | 4 | 1104 | |||
BC075655.1 | - | 3 | 847 | 30542293 |