Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271467.1 | NP_001258396.1 | 9 | 1282 | ||
NM_001271468.1 | NP_001258397.1 | 8 | 1187 | |||
NM_001271470.1 | NP_001258399.1 | 9 | 1349 | |||
NM_010249.2 | NP_034379.1 | 8 | 1187 | |||
NM_207669.2 | NP_997552.1 | 8 | 1184 | |||
GenBank mRNA | AI324106.1 | - | 1 | 266 | ||
AK052828.1 | - | 1163 | ||||
AK139710.1 | - | 9 | 1349 | |||
AK140222.1 | - | 9 | 1235 | |||
AK140907.1 | - | 8 | 1095 | |||
AK146373.1 | - | 8 | 1051 | |||
AW492609.1 | - | 1 | 271 | |||
AY282800.1 | - | 5 | 644 | |||
AY282803.1 | - | 5 | 644 | |||
AY282804.1 | - | 6 | 726 | |||
AY282805.1 | - | 5 | 727 | |||
BC013558.1 | - | 8 | 1078 | 3592119 | ||
M74516.1 | - | 8 | 1079 | |||
M74517.1 | - | 8 | 1081 |