Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF297615.1 | - | 7 | 739 | ||
AK004753.1 | - | 7 | 802 | |||
AK088605.1 | - | 6 | 537 | |||
AK159087.1 | - | 7 | 811 | |||
BC006810.1 | - | 5 | 492 | 3154188 | ||
M26925.1 | - | 6 | 583 | |||
M85153.1 | - | 7 | 682 |