Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010322.3 | NP_034452.3 | 10, 11 | 1722 | ||
GenBank mRNA | AF110769.1 | - | 10, 11 | 1686 | ||
AJ132012.1 | - | 10, 11 | 1517 | |||
AK004364.1 | - | 1, 2 | 169 | |||
AK004630.1 | - | 10, 11 | 1696 | |||
AK004771.1 | - | 10, 11 | 1702 | |||
AK010896.1 | - | 10, 11 | 1696 | |||
AK150810.1 | - | 10, 11 | 1680 | |||
AK166997.1 | - | 10, 11 | 1694 | |||
AK211963.1 | - | 2, 3 | 381 | |||
BC003225.1 | - | 10, 11 | 1664 | |||
BC025972.1 | - | 10, 11 | 1679 | 4020505 |