Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145130.2 | NP_660112.1 | 12 | 1530 | ||
GenBank mRNA | AB294194.1 | - | 12 | 1425 | ||
AK083687.1 | - | 10 | 1086 | |||
AK098090.1 | - | 12 | 1501 | |||
AK154062.1 | - | 12 | 1524 | |||
AK165319.1 | - | 12 | 1509 | |||
AK169452.1 | - | 11 | 1647 | |||
AK188090.1 | - | 2 | 206 | |||
AY028317.1 | - | 12 | 1506 | |||
BC006753.1 | - | 12 | 1458 | 3709076 |