Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008173.3 | NP_032199.3 | 1 | 1137 | ||
GenBank mRNA | AK037733.1 | - | 1 | 1017 | ||
AK155200.1 | - | 3 | 1219 | |||
AK156323.1 | - | 3 | 1213 | |||
BC008154.1 | - | 2 | 1445 | |||
BC016462.1 | - | 3 | 1220 | |||
BC129912.1 | - | 2 | 1115 | 40111802 | ||
BC129913.1 | - | 2 | 1115 | 40111803 | ||
DQ504162.1 | - | 1 | 1121 | |||
HM236293.1 | - | 2 | 1097 | |||
X04435.1 | - | 2 | 1115 | |||
X13358.1 | - | 2 | 1097 | |||
X13359.1 | - | 2 | 1097 |