Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159410.1 | NP_001152882.1 | 4, 5 | 285 | ||
NM_008193.3 | NP_032219.2 | 4, 5 | 448 | |||
GenBank mRNA | AK004205.1 | - | 4, 5 | 307 | ||
AK013514.1 | - | 4, 5 | 311 | |||
BC024625.1 | - | 4, 5 | 312 | 4485215 | ||
BY713424.1 | - | 4, 5 | 449 | |||
CO429095.1 | - | 4, 5 | 274 | |||
EL957688.1 | - | 4, 5 | 292 | |||
U53514.1 | - | 4, 5 | 268 |