Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020259.4 | NP_064655.4 | 9, 10 | 2037 | ||
GenBank mRNA | AF116865.1 | - | 9, 10 | 2018 | ||
AK031539.1 | - | 9, 10 | 2034 | |||
AK031841.1 | - | 9, 10 | 1547 | |||
BC053012.1 | - | 10, 11 | 2022 | 6402422 |