Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hivep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010437.2 | NP_034567.2 | 4 | 1495 | ||
GenBank mRNA | AK138961.1 | - | 2 | 369 | ||
AK143444.1 | - | 5 | 1305 | |||
AK147244.1 | - | 5 | 1576 | |||
AK147348.1 | - | 4 | 1495 | |||
AK147419.1 | - | 9 | 1785 | |||
AK147655.1 | - | 4 | 1495 | |||
BC150704.1 | - | 1 | 700 | 9087615 | ||
Y15907.1 | - | 1 | 746 |