Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146100.1 | NP_001139572.1 | 10, 11 | 1915 | ||
NM_010438.3 | NP_034568.2 | 14, 15 | 1858 | |||
GenBank mRNA | AK143630.1 | - | 10, 11 | 1853 | ||
AK149749.1 | - | 10, 11 | 1628 | |||
AK161466.1 | - | 4, 5 | 827 | |||
AK167468.1 | - | 10, 11 | 1671 | |||
AK185580.1 | - | 1, 2 | 96 | |||
AK199185.1 | - | 1, 2 | 96 | |||
BC027316.1 | - | 8, 9 | 1010 | |||
BC072628.1 | - | 10, 11 | 2036 | 6832742 | ||
J05277.1 | - | 10, 11 | 1573 | |||
L16949.1 | - | 14, 15 | 1837 |