Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301340.1 | NP_001288269.1 | 8 | 575 | ||
NM_001301341.1 | NP_001288270.1 | 10 | 702 | |||
NM_001301343.1 | NP_001288272.1 | 8 | 561 | |||
NM_001301344.1 | NP_001288273.1 | 7 | 503 | |||
NM_001301345.1 | NP_001288274.1 | 7 | 503 | |||
NM_025279.3 | NP_079555.1 | 8 | 575 | |||
GenBank mRNA | AK011428.1 | - | 8 | 580 | ||
AK051313.1 | - | 7 | 497 | |||
AK078777.1 | - | 10 | 703 | |||
AK088462.1 | - | 7 | 490 | |||
AK128891.1 | - | 8 | 568 | |||
AK135519.1 | - | 8 | 489 | |||
AK144245.1 | - | 7785 | ||||
AK150848.1 | - | 8 | 555 | |||
AK151689.1 | - | 7 | 457 | |||
AK152927.1 | - | 7 | 457 | |||
AK160615.1 | - | 8 | 545 | |||
AK160887.1 | - | 8 | 577 | |||
AK166656.1 | - | 8 | 580 | |||
AK166927.1 | - | 8 | 533 | |||
AK167600.1 | - | 8 | 566 | |||
AK168918.1 | - | 4 | 478 | |||
AK169064.1 | - | 8 | 565 | |||
AK191331.1 | - | 2 | 58 | |||
AK201722.1 | - | 2 | 58 | |||
AK204853.1 | - | 2 | 58 | |||
AK220023.1 | - | 2 | 58 | |||
BC006694.1 | - | 8 | 542 | 3583037 | ||
BC089328.1 | - | 8 | 498 | 6402614 | ||
EU604547.1 | - | 6 | 356 |