Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017371.2 | NP_059067.2 | 10 | 1273 | ||
GenBank mRNA | AA985967.1 | - | 1 | 213 | ||
AK145928.1 | - | 10 | 1271 | |||
BC011246.1 | - | 10 | 1253 | 4196684 | ||
BC019901.1 | - | 10 | 1255 | 5123914 | ||
U89889.1 | - | 10 | 1229 |