Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010485.3 | NP_034615.2 | 5 | 684 | ||
GenBank mRNA | AK034536.1 | - | 5 | 671 | ||
AK075818.1 | - | 5 | 646 | |||
AK078333.1 | - | 5 | 1000 | |||
AK080365.1 | - | 5 | 642 | |||
AK089088.1 | - | 4 | 391 | |||
AK148528.1 | - | 5 | 612 | |||
AK161667.1 | - | 4 | 401 | |||
AK201115.1 | - | 3 | 203 | |||
AK214281.1 | - | 3 | 229 | |||
U17595.1 | - | 2 | 99 | |||
U65735.1 | - | 4 | 453 |