Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252266.1 | NP_001239195.1 | 2 | 218 | ||
NM_010492.3 | NP_034622.3 | 3 | 631 | |||
GenBank mRNA | AB303567.1 | - | 2 | 142 | ||
AK005965.1 | - | 3 | 336 | |||
AK016989.1 | - | 3 | 334 | |||
AK192988.1 | - | 3 | 248 | |||
AK206264.1 | - | 11 | ||||
BC002030.1 | - | 3 | 265 | 3487814 | ||
BQ442697.1 | - | 3 | 631 | |||
CF914747.1 | - | 3 | 334 | |||
U26459.1 | - | 3 | 254 | |||
U26460.1 | - | 2 | 160 | |||
U26461.1 | - | 2 | 159 | |||
U34595.1 | - | 1 | 127 |