Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252266.1 | NP_001239195.1 | 2 | 176 | ||
NM_010492.3 | NP_034622.3 | 3 | 589 | |||
GenBank mRNA | AB303567.1 | - | 2 | 100 | ||
AK005965.1 | - | 3 | 294 | |||
AK016989.1 | - | 3 | 292 | |||
BC002030.1 | - | 3 | 223 | 3487814 | ||
CF914747.1 | - | 3 | 292 | |||
U26459.1 | - | 3 | 212 | |||
U26460.1 | - | 2 | 118 | |||
U26461.1 | - | 2 | 117 | |||
U34595.1 | - | 1 | 85 |